missing translation for 'onlineSavingsMsg'
Learn More

Potassium bicarbonate, Puriss., meets analytical spec. of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric), Honeywell Fluka™

Catalog No. p-7111749
Change view
Click to view available options
Quantity:
1 kg
6 x 1 kg
Packaging:
Poly bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
6001939 1 kg Poly bottle
60044737 6 x 1 kg Poly bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 6001939 Supplier Honeywell Chemicals Supplier No. 126021KG
Only null left

Puriss., meets analytical specification of BP, Ph. Eur., USP, E501, 99.5-101.0% (acidimetric)

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium;hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Quantity 1 kg
Linear Formula KHCO3
UN Number NONH for all modes of transport
Beilstein 4535309
Solubility Information Soluble in water
Formula Weight 100.12g/mol
Percent Purity 99.5 to 101% (acidimetric)
Grade puriss.
Packaging Poly bottle
Chemical Name or Material Potassium Bicarbonate, Meets analytical specification of Ph. Eur. BP, USP
EINECSNumber 206-059-0