missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Reagents
Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.
$71.07 - $213.20
Chemical Identifiers
| CAS | 298-14-6 |
|---|---|
| Molecular Formula | CHKO3 |
| Molecular Weight (g/mol) | 100.11 |
| MDL Number | MFCD00011402 |
| InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
| Synonym | Potassium Hydrogen Carbonate |
| IUPAC Name | potassium hydrogen carbonate |
| SMILES | [K+].OC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
01-337-125
|
Labchem Inc
C2277000100C2 |
100 g |
Each for $71.07
|
|
|||||
|
01-337-124
|
Labchem Inc
C2277000500A5 |
500 g |
Each for $213.20
|
|
|||||
Chemical Identifiers
| 298-14-6 | |
| 100.11 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| potassium hydrogen carbonate |
| CHKO3 | |
| MFCD00011402 | |
| Potassium Hydrogen Carbonate | |
| [K+].OC([O-])=O |
Specifications
| 127°C | |
| 298-14-6 | |
| 100 g | |
| MFCD00011402 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| potassium hydrogen carbonate | |
| Pure | |
| 2.17 |
| Colorless/White | |
| Solid | |
| CHKO3 | |
| Potassium Hydrogen Carbonate | |
| [K+].OC([O-])=O | |
| 100.11 | |
| Glass Bottle | |
| Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate) |