missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Reagents

Catalog No. 01337125
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337125 100 g
01337124 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01337125 Supplier Labchem Inc Supplier No. C2277000100C2

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate), Chemically Pure, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym Potassium Hydrogen Carbonate
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Melting Point 127°C
Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade Pure
Packaging Glass Bottle
Specific Gravity 2.17
Chemical Name or Material Potassium Bicarbonate, Purified (Potassium Hydrogen Carbonate)