missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Granular, USP, 99.5-101.5%, Spectrum™ Chemical

Catalog No. 18605460
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Poly Bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605460 45 kg Poly Bottle
18605457 500 g Poly Bottle
18605458 2.5 kg Poly Bottle
18605459 12 kg Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605460 Supplier Spectrum Chemical Mfg Cor Supplier No. PO15845KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

PO158, 298-14-6, KHCO3

Spectrum™ Chemical Potassium Bicarbonate, Granular, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 1
Physical Form Granular/Lump
Quantity 45 kg
Percent Purity 99.5 to 101.5%
Grade USP
Packaging Poly Bottle
Chemical Name or Material Potassium Bicarbonate, Granular