missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Granular, USP, 99.5-101.5%, Spectrum™ Chemical

PO158, 298-14-6, KHCO3

Supplier:  Spectrum Chemical Mfg Cor PO15812KG

 View more versions of this product

Catalog No. 18605459


Only null left
Explore available promotions

Description

Description

Spectrum™ Chemical Potassium Bicarbonate, Granular, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Specifications

Specifications

298-14-6
12 kg
MFCD00011402
[K+].OC([O-])=O
100.11
USP
1
CHKO3
TYJJADVDDVDEDZ-UHFFFAOYSA-M
potassium hydrogen carbonate
99.5 to 101.5%
Poly Pail

Chemical Identifiers

298-14-6
100.11
TYJJADVDDVDEDZ-UHFFFAOYSA-M
[K+].OC([O-])=O
CHKO3
MFCD00011402
potassium hydrogen carbonate
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Promotions

Promotions