missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate, Granular, USP, 99.5-101.5%, Spectrum™ Chemical
PO158, 298-14-6, KHCO3
Supplier: Spectrum Chemical Mfg Cor PO15825KG
Description
Spectrum™ Chemical Potassium Bicarbonate, Granular, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
298-14-6 | |
2.5 kg | |
MFCD00011402 | |
[K+].OC([O-])=O | |
100.11 | |
USP |
1 | |
CHKO3 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
potassium hydrogen carbonate | |
99.5 to 101.5% | |
Poly Bottle |
Chemical Identifiers
298-14-6 | |
100.11 | |
TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
[K+].OC([O-])=O |
CHKO3 | |
MFCD00011402 | |
potassium hydrogen carbonate |