missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Granular, USP, 99.5-101.5%, Spectrum™ Chemical

Catalog No. 18605457
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605457 500 g Poly Bottle
18605458 2.5 kg Poly Bottle
18605459 12 kg Poly Pail
18605460 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605457 Supplier Spectrum Chemical Mfg Cor Supplier No. PO158500GM

PO158, 298-14-6, KHCO3

Spectrum™ Chemical Potassium Bicarbonate, Granular, USP is a mineral supplement used to treat or prevent low amounts of potassium in the blood. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.5 to 101.5%
Grade USP
Packaging Poly Bottle