missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Granular, FCC, 99-101.5%, Spectrum™ Chemical

Catalog No. p-7158046
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605453 500 g Poly Bottle
18605454 2.5 kg Poly Pail
18605455 12 kg Poly Pail
18605456 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18605453 Supplier Spectrum Chemical Mfg Cor Supplier No. P1182500GM
Only null left

P1182, 298-14-6, KHCO3

Spectrum™ Chemical Potassium Bicarbonate, Granular, FCC is used as a carbon dioxide source for leavening in baking. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99 to 101.5%
Grade FCC
Loss on Drying 0.0025
Packaging Poly Bottle