missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, Crystal, ACS, 99.7-100.5%, Spectrum™ Chemical

Catalog No. p-7166033
Change view
Click to view available options
Quantity:
500 g
2.5 kg
12 kg
45 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612459 500 g Poly Bottle
18612460 2.5 kg Poly Bottle
18612461 12 kg Poly Pail
18612462 45 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18612459 Supplier Spectrum Chemical Mfg Cor Supplier No. P1175500GM
Only null left

Chemical Identifiers

CAS 298-14-6

Specifications

CAS Max % 1
Quantity 500 g
Percent Purity 99.7 to 100.5%
Grade ACS
Packaging Poly Bottle

CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.