missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate, Crystal, ACS, 99.7-100.5%, Spectrum™ Chemical
Specifications
Specifications
| CAS Max % | 1 |
| Physical Form | Crystals or Needles |
| Quantity | 45 kg |
| Molecular Weight (g/mol) | 100.12 |
| Percent Purity | 99.7 to 100.5% |
| Grade | ACS/Reagent |
| Packaging | Poly Bottle |
| Chemical Name or Material | Potassium Bicarbonate, Crystal |
CAUTION: For manufacturing or laboratory use only. Read and understand the label and Safety Data Sheet (SDS) prior to use.