missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals
$182.61 - $182.61
Chemical Identifiers
| CAS | 298-14-6 |
|---|---|
| Molecular Formula | CHKO3 |
| Molecular Weight (g/mol) | 100.11 |
| MDL Number | MFCD00011402 |
| InChI Key | TYJJADVDDVDEDZ-UHFFFAOYSA-M |
| Synonym | potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 |
| PubChem CID | 516893 |
| ChEBI | CHEBI:81862 |
| IUPAC Name | potassium hydrogen carbonate |
| SMILES | [K+].OC([O-])=O |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
ICN15255791
|
MP Biomedicals Inc
0215255791 |
1 kg |
Each for $182.61
|
|
|||||
Chemical Identifiers
| 298-14-6 | |
| 100.11 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| 516893 | |
| potassium hydrogen carbonate |
| CHKO3 | |
| MFCD00011402 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| CHEBI:81862 | |
| [K+].OC([O-])=O |
Specifications
| 298-14-6 | |
| 99.7-100.5% | |
| MFCD00011402 | |
| TYJJADVDDVDEDZ-UHFFFAOYSA-M | |
| potassium hydrogen carbonate | |
| 516893 | |
| 100.12 | |
| Potassium Bicarbonate |
| 1 kg | |
| CHKO3 | |
| potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3 | |
| [K+].OC([O-])=O | |
| 100.11 | |
| CHEBI:81862 | |
| ACS Reagent |
Safety and Handling
Recommended Storage : Room Temperature