missing translation for 'onlineSavingsMsg'
Learn More

Potassium Bicarbonate, 99.7-100.5%, (ACS Reagent Grade), MP Biomedicals

Catalog No. ICN15255791
Change view
Click to view available options
Quantity:
1 kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15255791 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN15255791 Supplier MP Biomedicals Inc Supplier No. 0215255791

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 298-14-6
Molecular Formula CHKO3
Molecular Weight (g/mol) 100.11
MDL Number MFCD00011402
InChI Key TYJJADVDDVDEDZ-UHFFFAOYSA-M
Synonym potassium bicarbonate, potassium hydrogen carbonate, potassium hydrogencarbonate, potassium acid carbonate, monopotassium carbonate, carbonic acid, monopotassium salt, unii-hm5z15lebn, potassiumbicarbonate, ccris 3510, khco3
PubChem CID 516893
ChEBI CHEBI:81862
IUPAC Name potassium hydrogen carbonate
SMILES [K+].OC([O-])=O

Specifications

Quantity 1 kg
Assay Percent Range 99.7-100.5%
Formula Weight 100.12
Grade ACS Reagent
Chemical Name or Material Potassium Bicarbonate
Recommended Storage Room Temperature