missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonyl tartrate trihydrate, ACS reagent

Catalog No. AC212801000
Change view
Click to view available options
Quantity:
100 g
Packaging:
Glass bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC212801000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC212801000 Supplier Thermo Scientific Chemicals Supplier No. 212801000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 28300-74-5 | 1·5 H2O | 333.93 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 28300-74-5
Molecular Formula 1·5 H2O
Molecular Weight (g/mol) 333.93
Synonym Antimonyl potassium tartrate sesquihydrate

Specifications

Melting Point 100.0°C
Packaging Glass bottle
Quantity 100 g
Merck Index 15691
Solubility Information Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water,1g/15mL glycerol,insoluble in alcohol
Formula Weight 333.93
Grade ACS Reagent
Chemical Name or Material Potassium antimonyl tartrate sesquihydrate
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful if inhaled.
Harmful if swallowed.
Precautionary Statement GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Avoid release to the environment.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

RUO – Research Use Only