missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium antimonyl tartrate trihydrate, 99+%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Melting Point | 100.0°C |
| Color | White |
| Physical Form | Fine Crystalline Powder |
| Quantity | 100 g |
| Assay Percent Range | 99% min. (Iodometry) |
| Merck Index | 15, 691 |
| Solubility Information | Solubility in water: 8.3%. Other solubilities: 1g/3mL boiling water, 1g/15mL glycerol, insoluble in alcohol |
| Formula Weight | 667.87 |
| Percent Purity | ≥99% |
| Infrared Spectrum | Authentic |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Toxic to aquatic life with long lasting effects. Harmful if inhaled. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician. Avoid release to the environment. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. |
| RTECSNumber | CC6825000 |
RUO – Research Use Only