missing translation for 'onlineSavingsMsg'
Learn More

Potassium Antimony Tartrate Trihydrate, ACS Reagent Grade, Ricca Chemical

Catalog No. RDCP0320100
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
RDCP0320100 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. RDCP0320100 Supplier Ricca Chemical Company Supplier No. RDCP0320100B1

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28300-74-5
Molecular Weight (g/mol) 671.9 g/mol

Specifications

CAS Min % 0.98
CAS Max % 1
Packaging White Poly Wide Mouth Bottle
Quantity 100 g
Percent Purity 99 to 103%
Grade ACS
Physical Form Crystalline Powder or Lumps
Chemical Name or Material Potassium Antimony Tartrate, Trihydrate
ShelfLife 60 months