missing translation for 'onlineSavingsMsg'
Learn More

Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic), Reagents

Catalog No. 01337482
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01337482 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01337482 Supplier Labchem Inc Supplier No. C2141550100C2
Only null left

Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 28300-74-5
Molecular Formula C8H10K2O15Sb2
Molecular Weight (g/mol) 667.87
InChI Key WBTCZEPSIIFINA-UHFFFAOYNA-J
Synonym Potassium Antimonyl Tartrate Trihydrate
IUPAC Name dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1⁴,⁷]tetradecane-3,9-dicarboxylate trihydrate
SMILES O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2

Specifications

Color Colorless/White
Physical Form Solid
Quantity 100 g
Grade ACS
Packaging Glass Bottle
Specific Gravity 2.607
Chemical Name or Material Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic)