missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic), Reagents
Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic), ACS, for general laboratory use. Manufactured in ISO 9001 facility.
Supplier: Labchem Inc C2141550500C2
Specifications
| Potassium Antimony Tartrate Trihydrate, ACS Reagent (Tartar Emetic) | |
| 28300-74-5 | |
| 500 g | |
| Potassium Antimonyl Tartrate Trihydrate | |
| O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2 | |
| 667.87 | |
| Glass Bottle |
| Colorless/White | |
| Solid | |
| C8H10K2O15Sb2 | |
| WBTCZEPSIIFINA-UHFFFAOYNA-J | |
| dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1â´,â·]tetradecane-3,9-dicarboxylate trihydrate | |
| ACS | |
| 2.607 |
Chemical Identifiers
| 28300-74-5 | |
| 667.87 | |
| Potassium Antimonyl Tartrate Trihydrate | |
| O.O.O.[K+].[K+].[O-]C(=O)C1O[Sb]2OC(C(O[Sb]3OC1C(=O)O3)C([O-])=O)C(=O)O2 |
| C8H10K2O15Sb2 | |
| WBTCZEPSIIFINA-UHFFFAOYNA-J | |
| dipotassium 5,11-dioxo-2,6,8,12,13,14-hexaoxa-1,7-distibatricyclo[8.2.1.1â´,â·]tetradecane-3,9-dicarboxylate trihydrate |