missing translation for 'onlineSavingsMsg'
Learn More

Potassium Antimony Tartrate Trihydrate, ACS, 99-103%, Spectrum™ Chemical

Catalog No. p-7165048
Change view
Click to view available options
Quantity:
25 g
125 g
500 g
2.5 kg
12 kg
50 kg
Packaging:
Fiber Drum
Poly Bottle
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18612450 25 g Poly Bottle
18612451 125 g Poly Bottle
18612452 500 g Poly Bottle
18612453 2.5 kg Poly Bottle
18612454 12 kg Poly Pail
18612455 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18612450 Supplier Spectrum Chemical Mfg Cor Supplier No. A156125GM
Only null left

Chemical Identifiers

CAS 28300-74-5

Specifications

CAS Max % 1
Loss on Drying 0.027
Packaging Poly Bottle
Quantity 25 g
Percent Purity 99 to 103%
Grade ACS

CAUTION: For manufacturing or laboratory use only. Not for food or drug use. Read and understand the label and Safety Data Sheet (SDS) prior to use.