missing translation for 'onlineSavingsMsg'
Learn More

Potassium Antimony Tartrate, ACS Grade, 99.0 to 103.0%, LabChem™

Catalog No. LC187109
Change view
Click to view available options
Quantity:
100 g
Packaging:
Poly Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
LC187109 100 g Poly Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. LC187109 Supplier LabChem Supplier No. LC187109

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 28300-74-5

Specifications

Acidity <0.020meq/gcSt
CAS Max % 1
Decomposition Information Carbon monoxide; Carbon dioxide; Antimony and its oxides
Color White
Density 2,600kg/m3
Packaging Poly Bottle
Linear Formula C8H4K2O12Sb2·3H2O
Quantity 100 g
UN Number UN1551
Specific Gravity 2600kg/m3
Solubility Information Soluble in water
Formula Weight 667.87
Percent Purity 99.0 to 103.0%
Grade ACS
Physical Form Powder
Chemical Name or Material Potassium Antimony Tartrate
Show More Show Less
Hazard Category Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Toxic to aquatic life.
Precautionary Statement GHS P Statement
Wash exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
Avoid release to the environment.
If swallowed: Rinse mouth.
Immediately call a poison center/doctor.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
EINECSNumber 234-293-3
Recommended Storage Room Temperature