missing translation for 'onlineSavingsMsg'
Learn More

Potassium antimonate trihydrate, 94+%

Catalog No. AA3952418
Change view
Click to view available options
Quantity:
50 g
250 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3952418 50 g
AA3952430 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3952418 Supplier Thermo Scientific Chemicals Supplier No. 03952418

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 12208-13-8 | KSbO3·3H2O

Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 12208-13-8
Molecular Formula KSbO3·3H2O
MDL Number MFCD00150439

Specifications

Color White
Physical Form Crystals or Powder
Quantity 50 g
Assay Percent Range >94%
UN Number 1549
Synonym Potassium Pyroantimonate, Potassium Hexahydroxyantimonate
Solubility Information Soluble in water and glycerine. Insoluble in ethanol.
Formula Weight 262.90 (208.85 Anhydrous)
Appearance White powder
Chemical Name or Material Potassium Antimonate Trihydrate
Hazard Category H302+H332
Precautionary Statement P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
DOTInformation DOT Class: 6.1, Packaging Group: III
EINECSNumber 235-387-7
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only