missing translation for 'onlineSavingsMsg'
Learn More

Potassium Alum, Dodecahydrate, Powder, USP, 99-100.5%, Spectrum™ Chemical

Catalog No. 18605440
Change view
Click to view available options
Quantity:
50 kg
Packaging:
Fiber Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18605440 50 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18605440 Supplier Spectrum Chemical Mfg Cor Supplier No. A273550KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

A2735, 7784-24-9, AlK(SO4)2·12H2O

Spectrum™ Chemical Potassium Alum, Dodecahydrate, Powder, USP is used as an antiseptic and astringent. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 7784-24-9
Molecular Formula AlH24KO20S2
Molecular Weight (g/mol) 474.37
InChI Key GNHOJBNSNUXZQA-UHFFFAOYSA-J
IUPAC Name aluminum(3+) potassium dodecahydrate disulfate
SMILES O.O.O.O.O.O.O.O.O.O.O.O.[Al+3].[K+].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O

Specifications

CAS Max % 1
Quantity 50 kg
Percent Purity 99 to 100.5%
Grade USP
Loss on Drying 43-46%
Packaging Fiber Drum