missing translation for 'onlineSavingsMsg'
Learn More

Potassium acetate hydrate, Puratronic™, 99.997% (metals basis)

CAS: 254966-93-3 | C2H3KO2 | 98.14 g/mol

$140.86 - $580.78

Chemical Identifiers

CAS 254966-93-3
Molecular Formula C2H3KO2
Molecular Weight (g/mol) 98.14
MDL Number MFCD00012458
InChI Key SCVFZCLFOSHCOH-UHFFFAOYSA-M
Synonym potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
PubChem CID 23702041
SMILES [K+].CC([O-])=O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA1297506
View Documents
Thermo Scientific Chemicals
01297506
5 g
Each for $140.86
Only null left
 
AA1297514
View Documents
Thermo Scientific Chemicals
01297514
25 g
Each for $580.78
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

254966-93-3
98.14
SCVFZCLFOSHCOH-UHFFFAOYSA-M
23702041
C2H3KO2
MFCD00012458
potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
[K+].CC([O-])=O

Specifications

White
Crystalline
C2H3KO2
potassium acetate hydrate, c2h3o2.k.h2o, potassium acetate hydrate, puratronic, potassium acetate hydrate, puratronic r metals basis
[K+].CC([O-])=O
98.14
98.15 (Anhydrous)
Puratronic
(metals basis)
254966-93-3
5 g
MFCD00012458
SCVFZCLFOSHCOH-UHFFFAOYSA-M
potassium acetate
23702041
99.997%
Potassium acetate hydrate
Videos
Safety and Handling

Safety and Handling

EINECSNumber : 204-822-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'

RUO – Research Use Only