missing translation for 'onlineSavingsMsg'
Learn More

Potassium, 98%, morceaux, dans l’huile minérale

Numéro de catalogue. AC223865000
Modifier l'affichage
Click to view available options
Quantity:
10 g
50 g
100 g
500 g
Packaging:
Bouteille en plastique
Bouteille en plastique dans une boîte de conserve
Fût en plastique
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Numéro de catalogue. Quantity Packaging
AC223865000 500 g Fût en plastique
AC223860100 10 g Bouteille en plastique dans une boîte de conserve
AC223860500 50 g Bouteille en plastique
AC223861000 100 g Bouteille en plastique
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Numéro de catalogue. AC223865000 Fournisseur Thermo Scientific Chemicals Code fournisseur 223865000

CAS : 7440-09-7 | K | 39,10 g/mol

Ce produit de marque Thermo Scientific Chemicals faisait à l’origine partie du portefeuille Acros Organics. Certains documents et informations d’étiquetage peuvent faire référence à l’ancienne marque. Le code produit/article (SKU) original d’Acros Organics n’a pas changé dans le cadre de la transition vers la marque Thermo Scientific Chemicals.

Identifiants chimiques

CAS 9-7-7440
Molecular Formula K
Molecular Weight (g/mol) 39.10
MDL Number MFCD00133776
InChI Key ZLMJMSJWJFRBEC-UHFFFAOYSA-N
Synonym kalium, potassium, metal, potassium, liquid alloy, unii-rwp5ga015d, rwp5ga015d, hsdb 698, monopotassium, potasio, mono-potassium, atom
PubChem CID 5462222
ChEBI CHEBI:26216
IUPAC Name potassium
SMILES [K]

Spécifications

Color Gris
Quantity 500 g
Packaging Fût en plastique
Form Chunk
Assay Percent Range 98%
Linear Formula K
Fieser 01,905; 06,274; 12,400; 13,170; 14,308; 15,269
Merck Index 15, 7725
Formula Weight 39.1
Chemical Name or Material Potassium
Hazard Category Danger
Hazard Statement GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
In contact with water releases flammable gases which may ignite spontaneously.
Reacts violently with water.
Precautionary Statement GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Usage réservé à la recherche