missing translation for 'onlineSavingsMsg'
Learn More

Potassium 5-Methyl-1,3,4-oxadiazole-2-carboxylate 98.0+%, TCI America™

Catalog No. p22201g
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
1 g
200 mg
Catalog No. Quantity
P22201G 1 g
P2220200MG 200 mg
2 options

Catalog No. P22201G

Supplier: TCI America P22201G

Only null left

Chemical Identifiers

CAS 888504-28-7
Molecular Formula C4H3KN2O3
Molecular Weight (g/mol) 166.177
MDL Number MFCD08060085
InChI Key BRHMYHXNUGVBCU-UHFFFAOYSA-M
Synonym 5-Methyl-1,3,4-oxadiazole-2-carboxylic Acid Potassium Salt
PubChem CID 24820502
IUPAC Name potassium;5-methyl-1,3,4-oxadiazole-2-carboxylate
SMILES CC1=NN=C(O1)C(=O)[O-].[K+]

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 166.18
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 5-Methyl-1,3,4-oxadiazole-2-carboxylate