missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Potassium 4-Pyridyltrifluoroborate 98.0+%, TCI America™
$109.98 - $441.78
Chemical Identifiers
| CAS | 1111732-87-6 |
|---|---|
| Molecular Formula | C5H4BF3KN |
| Molecular Weight (g/mol) | 184.998 |
| InChI Key | SLPOSOZRTRIRQS-UHFFFAOYSA-N |
| PubChem CID | 25212881 |
| IUPAC Name | potassium;trifluoro(pyridin-4-yl)boranuide |
| SMILES | [B-](C1=CC=NC=C1)(F)(F)F.[K+] |
Chemical Identifiers
| 1111732-87-6 | |
| 184.998 | |
| 25212881 | |
| [B-](C1=CC=NC=C1)(F)(F)F.[K+] |
| C5H4BF3KN | |
| SLPOSOZRTRIRQS-UHFFFAOYSA-N | |
| potassium;trifluoro(pyridin-4-yl)boranuide |
Specifications
| 1111732-87-6 | |
| C5H4BF3KN | |
| SLPOSOZRTRIRQS-UHFFFAOYSA-N | |
| potassium;trifluoro(pyridin-4-yl)boranuide | |
| 25212881 | |
| ≥98.0% (T) | |
| Potassium 4-Pyridyltrifluoroborate |
| White-Yellow | |
| 1 g | |
| [B-](C1=CC=NC=C1)(F)(F)F.[K+] | |
| 184.998 | |
| 185.00 | |
| Crystalline Powder |
Safety and Handling
TSCA : No