missing translation for 'onlineSavingsMsg'
Learn More

Potassium 4-Pyridyltrifluoroborate 98.0+%, TCI America™

Catalog No. P16845G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P16845G 5 g
P16841G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P16845G Supplier TCI America Supplier No. P16845G

Chemical Identifiers

CAS 1111732-87-6
Molecular Formula C5H4BF3KN
Molecular Weight (g/mol) 184.998
InChI Key SLPOSOZRTRIRQS-UHFFFAOYSA-N
PubChem CID 25212881
IUPAC Name potassium;trifluoro(pyridin-4-yl)boranuide
SMILES [B-](C1=CC=NC=C1)(F)(F)F.[K+]

Specifications

Color White-Yellow
Quantity 5 g
Formula Weight 185.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 4-Pyridyltrifluoroborate