missing translation for 'onlineSavingsMsg'
Learn More

Potassium (4-Methoxyphenyl)trifluoroborate 98.0+%, TCI America™

Catalog No. p-7135272
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18201G 1 g
P18205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18201G Supplier TCI America Supplier No. P18201G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 192863-36-8
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.036
MDL Number MFCD04112717
InChI Key XNYMCUFKZHRYID-UHFFFAOYSA-N
Synonym potassium trifluoro 4-methoxyphenyl borate, potassium 4-methoxyphenyl trifluoroborate, potassium 4-methoxyphenyltrifluoroborate, potassium trifluoro 4-methoxyphenyl boranuide, pubchem11347, amtb103, potassum 4-methoxyphenyl trfluoroborate, trifluoro 4-methoxyphenyl potassioboron v, potassium trifluoro-4-methoxyphenyl boranuide
PubChem CID 23683762
IUPAC Name potassium;trifluoro-(4-methoxyphenyl)boranuide
SMILES [B-](C1=CC=C(C=C1)OC)(F)(F)F.[K+]

Specifications

Color White
Physical Form Crystalline Powder
Quantity 1 g
Formula Weight 214.04
Percent Purity ≥98.0% (HPLC,T)
Chemical Name or Material Potassium (4-Methoxyphenyl)trifluoroborate