missing translation for 'onlineSavingsMsg'
Learn More

Potassium (3-Nitrophenyl)trifluoroborate 98.0+%, TCI America™

Catalog No. P18191G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18191G 1 g
P18195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18191G Supplier TCI America Supplier No. P18191G

Chemical Identifiers

CAS 192863-40-4
Molecular Formula C6H4BF3KNO2
Molecular Weight (g/mol) 229.01
MDL Number MFCD03701627
InChI Key ZKZMSCLFLMORNI-UHFFFAOYSA-N
Synonym potassium 3-nitrophenyl trifluoroborate, potassium 3-nitrophenyltrifluoroborate, potassium trifluoro 3-nitrophenyl boranuide, potassium trifluoro 3-nitrophenyl borate, potassium trifluoro-3-nitrophenyl boranuide, pubchem11373, potassium ion trifluoro 3-nitrophenyl boranuide, 3-nitrophenyltrifluoroborate potassium salt, potassium tris fluoranyl-3-nitrophenyl boranuide
PubChem CID 2782884
IUPAC Name potassium trifluoro(3-nitrophenyl)boranuide
SMILES [K+].[O-][N+](=O)C1=CC=CC(=C1)[B-](F)(F)F

Specifications

Melting Point 260°C
Color White
Quantity 1 g
Formula Weight 229.01
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (3-Nitrophenyl)trifluoroborate