missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-Indoleacetate 98.0+%, TCI America™

Catalog No. I002325G
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I002325G 25 g
I00231G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I002325G Supplier TCI America Supplier No. I002325G

Chemical Identifiers

CAS 2338-19-4
Molecular Formula C10H8KNO2
Molecular Weight (g/mol) 213.28
MDL Number MFCD00066213
InChI Key MLWMEUAQPRACHM-UHFFFAOYSA-M
Synonym Heteroauxine, 3-Indoleacetic Acid Potassium Salt, K-IAA
PubChem CID 23667427
IUPAC Name potassium 2-(1H-indol-3-yl)acetate
SMILES [K+].[O-]C(=O)CC1=CNC2=CC=CC=C12

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 213.28
Percent Purity ≥98.0% (HPLC,N)
Physical Form Crystalline Powder
Chemical Name or Material Potassium 3-Indoleacetate
EINECSNumber (5)-0132
TSCA No