missing translation for 'onlineSavingsMsg'
Learn More

Potassium 3-cyanophenyltrifluoroborate, 95%, Thermo Scientific™

$80.85 - $80.85

Chemical Identifiers

CAS 850623-46-0
Molecular Formula C7H4BF3KN
Molecular Weight (g/mol) 209.02
MDL Number MFCD04115744
InChI Key STQLGUGARUCKLI-UHFFFAOYSA-N
Synonym potassium 3-cyanophenyl trifluoroborate, potassium 3-cyanophenyltrifluoroborate, potassium 3-cyanophenyl trifluoroboranuide, amtb705, trifluoro 3-cyanophenyl potassioboron v, potassium 3-cyanophenyl-trifluoroboranuide, potassium ion 3-cyanophenyl trifluoroboranuide, potassium 3-cyanophenyl trifluoro borate 1-, potassium 3-cyanophenyl-tris fluoranyl boranuide
PubChem CID 23716868
SMILES [K+].F[B-](F)(F)C1=CC(=CC=C1)C#N
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH5267103
View Documents
Thermo Scientific Chemicals
H5267103
1 g N/A
Only null left
 
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

850623-46-0
209.02
STQLGUGARUCKLI-UHFFFAOYSA-N
23716868
C7H4BF3KN
MFCD04115744
potassium 3-cyanophenyl trifluoroborate, potassium 3-cyanophenyltrifluoroborate, potassium 3-cyanophenyl trifluoroboranuide, amtb705, trifluoro 3-cyanophenyl potassioboron v, potassium 3-cyanophenyl-trifluoroboranuide, potassium ion 3-cyanophenyl trifluoroboranuide, potassium 3-cyanophenyl trifluoro borate 1-, potassium 3-cyanophenyl-tris fluoranyl boranuide
[K+].F[B-](F)(F)C1=CC(=CC=C1)C#N

Specifications

850623-46-0
C7H4BF3KN
1 g
STQLGUGARUCKLI-UHFFFAOYSA-N
potassium (3-cyanophenyl)trifluoroboranuide
23716868
95%
196°C to 200°C
MFCD04115744
potassium 3-cyanophenyl trifluoroborate, potassium 3-cyanophenyltrifluoroborate, potassium 3-cyanophenyl trifluoroboranuide, amtb705, trifluoro 3-cyanophenyl potassioboron v, potassium 3-cyanophenyl-trifluoroboranuide, potassium ion 3-cyanophenyl trifluoroboranuide, potassium 3-cyanophenyl trifluoro borate 1-, potassium 3-cyanophenyl-tris fluoranyl boranuide
[K+].F[B-](F)(F)C1=CC(=CC=C1)C#N
209.02
209.02
Potassium 3-cyanophenyltrifluoroborate
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

Recommended Storage : Ambient temperatures

SDS
missing translation for 'documents'

missing translation for 'documents'