missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-naphthalenetrifluoroborate, 96%, Thermo Scientific™

Catalog No. p-7035187
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH5290103 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH5290103 Supplier Thermo Scientific Chemicals Supplier No. H5290103

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 668984-08-5
Molecular Formula C10H7BF3K
Molecular Weight (g/mol) 234.07
MDL Number MFCD06659941
InChI Key FXRJHJJHIOIXKE-UHFFFAOYSA-N
Synonym potassium 2-naphthalenetrifluoroborate, potassium trifluoro naphthalen-2-yl boranuide, potassium trifluoro naphthalen-2-yl borate, potassium 2-naphthalene trifluoroborate, pubchem11526, trifluoro 2-naphthyl potassioboron v, potassium trifluoro 2-naphthalenyl boranuide, potassium trifluoro 2-naphthyl borate, potassium trifluoro naphthalen-2-yl borate 1-, potassium ion trifluoro naphthalen-2-yl boranuide
PubChem CID 23674705
IUPAC Name potassium;trifluoro(naphthalen-2-yl)boranuide
SMILES [B-](C1=CC2=CC=CC=C2C=C1)(F)(F)F.[K+]

Specifications

Melting Point >300°C
Quantity 1 g
Formula Weight 234.07
Percent Purity 96%
Chemical Name or Material Potassium 2-naphthalenetrifluoroborate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures