missing translation for 'onlineSavingsMsg'
Learn More

Potassium (2-Methoxyphenyl)trifluoroborate 98.0+%, TCI America™

Catalog No. P18105G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P18105G 5 g
P18101G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P18105G Supplier TCI America Supplier No. P18105G

Chemical Identifiers

CAS 236388-46-8
Molecular Formula C7H7BF3KO
Molecular Weight (g/mol) 214.04
MDL Number MFCD04115756
InChI Key IXJPOOXJHKETAZ-UHFFFAOYSA-N
Synonym potassium 2-methoxyphenyl trifluoroborate, potassium trifluoro 2-methoxyphenyl borate, potassium 2-methoxyphenyltrifluoroborate, potassium trifluoro 2-methoxyphenyl boranuide, pubchem11466, amtb101, trifluoro 2-methoxyphenyl potassioboron v, potassium trifluoro-2-methoxyphenyl boranuide, potassium trifluoro 2-methoxyphenyl borate 1-
PubChem CID 23684966
IUPAC Name potassium trifluoro(2-methoxyphenyl)boranuide
SMILES [K+].COC1=CC=CC=C1[B-](F)(F)F

Specifications

Color White
Quantity 5 g
Formula Weight 214.04
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Potassium (2-Methoxyphenyl)trifluoroborate