missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-Ethylhexanoate 95.0+%, TCI America™

Catalog No. p-7128848
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E012125G 25 g
E0121500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E012125G Supplier TCI America Supplier No. E012125G
Only null left

Chemical Identifiers

CAS 3164-85-0
Molecular Formula C8H15KO2
Molecular Weight (g/mol) 182.30
MDL Number MFCD00045896
InChI Key ZUFQCVZBBNZMKD-UHFFFAOYNA-M
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
PubChem CID 23669619
IUPAC Name potassium 2-ethylhexanoate
SMILES [K+].CCCCC(CC)C([O-])=O

Specifications

Color White
Quantity 25 g
Formula Weight 182.30
Percent Purity ≥95.0% (T)
Physical Form Crystalline Lumps
Chemical Name or Material Potassium 2-Ethylhexanoate
EINECSNumber (2)-0611
TSCA Yes