missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-ethylhexanoate hydrate, 95%

Catalog No. p-7050776
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3944006 5 g
AA3944014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3944006 Supplier Thermo Scientific Chemicals Supplier No. 03944006

CAS: 3164-85-0 | C8H15KO2 | 182.30 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3164-85-0
Molecular Formula C8H15KO2
Molecular Weight (g/mol) 182.30
MDL Number MFCD00045896
InChI Key ZUFQCVZBBNZMKD-UHFFFAOYNA-M
PubChem CID 23669619
SMILES [K+].CCCCC(CC)C([O-])=O

Specifications

Odor Odorless
Linear Formula K[OOCCH(C2H5)C4H9]·xH2O
Quantity 5 g
Sensitivity Moisture sensitive
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
IUPAC Name potassium;2-ethylhexanoate
Formula Weight 182.30 (Anhydrous)
Percent Purity 95%
Physical Form Lump or Semiliquid
Chemical Name or Material Potassium 2-ethylhexanoate hydrate
Hazard Category H315-H318-H361
Hazard Statement GHS H Statement
H361
Suspected of damaging fertility or the unborn child.
Precautionary Statement P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
EINECSNumber 221-625-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only