missing translation for 'onlineSavingsMsg'
Learn More

Potassium 2-ethylhexanoate, 99.9% (metals basis), 75% w/w soln.

Catalog No. AA3916318
Change view
Click to view available options
Quantity:
2 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA3916318 50 g
AA3916304 2 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA3916318 Supplier Thermo Scientific Chemicals Supplier No. 03916318

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 3164-85-0 | C8H15KO2 | 182.30 g/mol

Used to convert the tert-butylammmonium salt of clavulanic acid into potassium clavulanate or clavulanate potassium. Also used as a catalyst for polyurethane systems (foams) and for unsaturated polyester resin systems.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 3164-85-0
Molecular Formula C8H15KO2
Molecular Weight (g/mol) 182.30
MDL Number MFCD00045896
InChI Key ZUFQCVZBBNZMKD-UHFFFAOYNA-M
PubChem CID 23669619
SMILES [K+].CCCCC(CC)C([O-])=O

Specifications

Odor Characteristic
Linear Formula K[OOCCH(C2H5)C4H9]
Quantity 50 g
UN Number UN1993
Synonym potassium 2-ethylhexanoate, hexanoic acid, 2-ethyl-, potassium salt, 2-ethylhexanoic acid potassium salt, 2-ethylhexanoic acid, potassium salt, hexanoic acid, 2-ethyl-, potassium salt 1:1, potassium2-ethylhexanoate, dsstox_cid_7525, dsstox_rid_78488, dsstox_gsid_27525, ksc493c3p
Solubility Information Partly miscible with water.
IUPAC Name potassium;2-ethylhexanoate
Formula Weight 182.3
Percent Purity 99.9%
Concentration or Composition (by Analyte or Components) 75% w/w soln.
Physical Form Viscous Liquid
Assay (metals basis)
Chemical Name or Material Potassium 2-ethylhexanoate
Show More Show Less
Hazard Category H315-H318-H361
Hazard Statement GHS H Statement
H226-H361-H373
Flammable liquid and vapor.
Suspected of damaging fertility or the unborn child.
May cause damage to organs through prolonged or repeated exposure.
Precautionary Statement P201-P202-P264b-P281-P302+P352-P305+P351+P338-P308+P313-P310-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 221-625-7
TSCA Yes
Recommended Storage Ambient temperatures

For Manufacturing and Laboratory Use. Not for beverage use