missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Positive Displacement Pipette Tips
Contains replacement consumables to run all Saftest™ test kits.
Supplier: MP Biomedicals™ 07DP1038
Description
- For use with SafTest analyzer
- Used with all six SafTest test kits
- Appropriate for all testing endpoints
- Research area: Lipid degradation and oxidation
- Detection method: Bichromatic photometer
- Application area: Oil/fat degradation
- Construction: Polystryene
- Pre assembled, pre sterilized
Specifications
| apillaries and pistons are pre-assembled and presterilized | |
| Sterile | |
| Polypropylene |
| 200 to 1000 μL | |
| SafTest Analyzer |