Learn More
ICU Medical Portex™ Line Draw Plus Arterial Blood Sampling Kits
Line Draw Plus aspirating syringes allow for blood sampling directly from an arterial line. The syringes use an advanced formula of dry lithium heparin for consistent and accurate readings. They are available with the following configurations: 1 mL Luer Slip Syringe, 3 mL Luer Lock Syringe, or 3 mL Luer Slip Slip Syringe.
$975.27 - $1775.07
| Catalog Number | Mfr. No. | Quantity | Volume (Metric) | Packaging | Product Type | Price | Quantity | ||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Volume (Metric) | Packaging | Product Type | Price | Quantity | ||
22-004-197
|
ICU Medical
40412 |
200/Cs. | 1 mL | Polybag | Lithium Heparin LS Syringe |
Case of 200 Each for $1,174.37
|
|
||
22-024-978
|
ICU Medical
4041LH |
200/Cs. | 1 mL | Polybag | Low Lithium Heparin LS Syringe |
Case of 200 Each for $975.27
|
|
||
22-024-957
|
ICU Medical
4041W2 |
200/Cs. | 1 mL | Polybag | Li Heparin Syringe, Waste Syr |
Case of 200 Each for $1,775.07
|
|
||
22-024-958
|
ICU Medical
4042LH |
200/Cs. | 3 mL | Polybag | Low Lithium Heparin LL Syringe |
Case of 200 Each for $1,032.65
|
|
||
Description
Advanced formula offsets both binding and dilution factors by artificially pre-adjusting each electrolyte and metabolite to its normal blood level. Provides consistent and accurate readings for pH, pCO2, pO2, Ca2+ , Mg2+ , Na+, K+, Cl-, CO-oximetry, glucose, and lactate.
- Umbilical and arterial syringes contain a 23.5 IU/mL concentration of dry lithium heparin
- Low-heparin arterial syringes have a 7 IU/mL concentration of dry lithium heparin
- Anaerobic syringes contain 23.5 IU/mL of electrolyte-balanced liquid sodium heparin
†With support kit (alcohol prep pad, gauze pad, biohazard-labeled ice bag).
Specifications
| 200/Cs. |