missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Port Plate with 3/8 in. Barbed Flange - E8402 - QC

Catalog No. 01000087
Change view
Click to view available options
Quantity:
100 Pk.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01000087 100 Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01000087

Ribbed base prevents suction blockage and increases rigidity. Bag ports, also known as port plates and port discs, are designed to be ultrasonic or thermal welded to medical and bio-reactor bags of like material. These single-use bags are used to transfer drug compounds, cell culture and fermentation as well as other media and fluids in the bio-pharmaceutical industry.

  • Single & Multi Use Applications
  • PVC-Free
  • Superior Single Barb Design
  • USP Class VI Compliant
  • RoHS Compliant
  • Gamma Stable
  • Animal Derivative Free
  • Lot Traceable

Warranty

Replacement if not satisfied. 12 Month return Policy.

Specifications

Certifications/Compliance Compliance Certificates Available
Description Class 7 Cleanroom Manufactured
Material Engage E8402
For Use With (Application) Single-use pharmaceutical bio-reactor bag applications. Biomedical and laboratory environment in fluid management applications.
Storage Requirements Out of direct sunlight, >20°F <100°F
Color Natural
Product Type Bag Port
Quantity 100 Pk.