International Crystal Lab KBr Port-A-Press™ Kit 7mm Deset
Catalog No. NC9256144
KBr Port-A- Press kit, complete with 7mm die set, torque wrench, channel holder and pellet press, ICL's Port-A-Press is the most economical, full-scale, it produces pellets are transparent as those made with the 12 ton hydraulic press
Catalog No.
NC9256144
Supplier
International Crystal LabSupplier No.
0012188
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
KBr Port-A- Press kit, complete with 7mm die set, torque wrench, channel holder and pellet press, ICL's Port-A-Press is the most economical, full-scale, it produces pellets are transparent as those made with the 12 ton hydraulic press