missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ AAV9 Column

Catalog No. A65885
Change view
Click to view available options
Diameter (Metric) Inner:
2.5 cm
5 cm
8 cm
10 cm
Length (Metric):
5 cm
10 cm
Column Capacity:
25 mL
50 mL
100 mL
200 mL
250 mL
390 mL
500 mL
785 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Length (Metric) Column Capacity
A65885 10 cm 10 cm 785 mL
A65872 2.5 cm 5 cm 25 mL
A65873 2.5 cm 10 cm 50 mL
A65880 5 cm 5 cm 100 mL
A65881 5 cm 10 cm 200 mL
A65882 8 cm 5 cm 250 mL
A65883 8 cm 10 cm 500 mL
A65884 10 cm 5 cm 390 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A65885 Supplier Thermo Scientific™ Supplier No. A65885

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAV9 affinity resin, POROS GoPure AAV9 columns offer high specificity and capacity for AAV9 viral vectors over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2–30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAV9 resin include one-step AAV9 purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAV9 ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

Specifications

Diameter (Metric) Inner 10 cm
Length (English) 4 in.
Quantity 1 Ea.
Type Chromatography Column
Pore Size Not Applicable
Column Capacity 785 mL
Length (Metric) 10 cm

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.