missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ GoPure™ AAV9 Column

Catalog No. A65872
Change view
Click to view available options
Diameter (Metric) Inner:
2.5 cm
5 cm
8 cm
10 cm
Length (Metric):
5 cm
10 cm
Column Capacity:
25 mL
50 mL
100 mL
200 mL
250 mL
390 mL
500 mL
785 mL
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Inner Length (Metric) Column Capacity
A65872 2.5 cm 5 cm 25 mL
A65873 2.5 cm 10 cm 50 mL
A65880 5 cm 5 cm 100 mL
A65881 5 cm 10 cm 200 mL
A65882 8 cm 5 cm 250 mL
A65883 8 cm 10 cm 500 mL
A65884 10 cm 5 cm 390 mL
A65885 10 cm 10 cm 785 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. A65872 Supplier Thermo Scientific™ Supplier No. A65872

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards.

POROS GoPure AAV9 chromatography columns are ready-to-use and designed specifically for use in clinical and commercial manufacturing. POROS GoPure AAV9 columns are prepared and packed in an ISO Class 7 environment where all product contact materials meet USP <88> Class VI standards. Each column is accompanied by column qualification documentation including bioburden and endotoxin. Packed with POROS™ CaptureSelect™ AAV9 affinity resin, POROS GoPure AAV9 columns offer high specificity and capacity for AAV9 viral vectors over a wide range of flow rates.

  • Pressure rating (bar/psi): 4/58
  • Operating temperature: 2–30°C
  • Bed support size: 12 μm
  • Inlet/outlet internal diameter: 2.00 mm (2.5 cm), 3.45 mm (5 cm), 4.57 mm (8 cm), 6.35 mm (10 cm)
  • Inlet/outlet port connections: 3/4 inch mini tri-clamps
  • Includes a 30 mL resin side sample for incoming QC

Features of POROS CaptureSelect AAV9 resin include one-step AAV9 purification from crude material with high purity, with high specificity and capacity, improving yield by reducing process steps. Free of animal-origin components, the CaptureSelect AAV9 ligand is a 12 to 15 kDa single-domain antibody fragment produced in the yeast Saccharomyces cerevisiae.

Specifications

Quantity 1 Ea.
Length (English) 2 in.
Type Chromatography Column
Diameter (Metric) Inner 2.5 cm
Pore Size Not Applicable
Column Capacity 25 mL
Length (Metric) 5 cm

For research and development use and further manufacturing in support of FDA-regulated end uses. Not for diagnostic use or direct administration to human or animals.