missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ Caprylate RoboColumn™ Pre-packed columns
GREENER_CHOICE

Catalog No. A51056
Change view
Click to view available options
Bed Volume:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
A51056 200 μL
A51057 600 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A51056 Supplier Thermo Scientific™ Supplier No. A51056

These Thermo Scientific™ POROS™ Caprylate RoboColumn™ are pre-packed with POROS Caprylate Mixed-Mode Chromatography Resin to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode.

These RoboColumns are pre-packed with POROS Caprylate Mixed-Mode Chromatography Resin to remove moderate to high levels of aggregates during downstream processing of therapeutic antibodies in flow-through mode. As a mixed-mode, weak cation-exchange resin, POROS Caprylate resin effectively removes other impurities, such as host cell proteins and a wide range of high molecular weight species. RoboColumns are small chromatography columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.

Features of POROS Caprylate Mixed-Mode resin include:

  • Selective capture of aggregates and hydrophobic high-molecular-weight impurities
  • Flow-through design that helps save buffer and material costs
  • Excellent scalability and high productivity
  • Wide operational range
  • Non-animal-derived

POROS Caprylate Mixed-Mode resin (Cat. No. A51051) operates in flow-through mode, which helps reduce costs, especially when working with more engineered monoclonal antibody derivatives. The unique selectivity of the caprylate ligand contributes to a high yield of the targeted monomer while using established cation exchange resin buffer systems. POROS Caprylate Mixed-Mode resin is manufactured using a synthetic manufacturing process that is free of animal components.

Main characteristics

  • Matrix: cross-linked poly(styrene-divinylbenzene) POROS beads
  • Average particle size: 50 μm
  • Ligand: octanoic acid
  • Binding capacity: >20 g lysozyme/L resin
  • Typical buffers: typical cation exchange buffers, pH 4.5–7.0, conductivity 10–30 mS/cm
  • Clean-in-place: 0.5 M NaOH
  • Formulation buffer: 0.1 M NaOH
  • Mechanical resistance: 100 bar (10 MPa, 1450 psi)

Other POROS chromatography resins are also available in this column format. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

RoboColumn™ is a registered trademark of ATOLL GmbH.

Specifications

Max. Pressure 8 bar
Pore Size Not Applicable
Stationary Phase Caprylic Acid
Length (Metric) 1 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm (nominal)
For Use With (Equipment) Robotic Fluid Handlers
Type Pre-packed Column
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 30°C.
Product Line POROS RoboColumn
Ligand Type Caprylic Acid
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Target Monoclonal Antibodies
Column Format Pre-packed Robo Column
Bed Volume 200 μL
Show More Show Less