missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ POROS™ AAVX RoboColumn™ Pre-packed columns
GREENER_CHOICE

Catalog No. A37795
Change view
Click to view available options
Bed Volume:
200 μL
600 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Bed Volume
A37795 600 μL
A37794 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A37795 Supplier Thermo Scientific™ Supplier No. A37795

Thermo Scientific™ RoboColumn™ chromatography products are small columns provided in 8-column strips.

RoboColumn™ chromatography products are small columns provided in 8-column strips. They are useful for fully automated and parallel chromatographic separations using robotic liquid handling platforms.

These RoboColumn™ columns are pre-packed with POROS CaptureSelect AAVX Affinity Resin (Cat. No. A36743), engineered to address the high selectivity and capacity requirements for downstream purification of a broad range of naturally occurring and synthetic adeno-associated virus (AAV) serotypes used for gene therapy applications. POROS CaptureSelect AAVX resin has demonstrated binding reactivity towards a set of AAV serotypes that includes AAV1 to AAV8 and AAVrh10, as well as synthetic serotypes.

POROS CaptureSelect AAVX resin enables a robust, fast, and platformable purification process for wide-ranging AAV serotypes with excellent purity and yield obtained in one step.

Features of POROS CaptureSelect AAVX resin include:

  • One-step AAV purification from crude material with high purity and yield
  • High specificity and capacity, maximizing yield by significantly reducing process steps
  • Superior capacity maintained at high flow rates
  • Excellent scalability and easier handling and packing due to rigid polymeric bead
  • Non-animal derived

Free of animal components

CaptureSelect products contain affinity ligands based on recombinant single-domain antibody fragments (VHH) created by a proprietary technology. The VHH affinity ligand is a 12 to 15 kDa fragment comprising the three complementarity-determining regions (CDRs) that form the antigen-binding domain, efficiently produced in the yeast Saccharomyces cerevisiae by a production process free of any animal components (Animal Origin-Free).

A wide selection of RoboColumn™ is available, including columns pre-packed with POROS ion exchange, HIC, and CaptureSelect affinity resins. All POROS high performance chromatography resins have a unique pore structure that provides excellent mass transport for high capacity and resolution over a wide range of flow rates.

RoboColumn™ is a registered trademark of Repligen Corporation.

Specifications

Max. Pressure 8 bar
Pore Size Not Applicable
Stationary Phase Immobilized CaptureSelect™ AAVX Ligand
Column Capacity 600 μL
Length (Metric) 3 cm
Quantity 1 Strip, 8 Columns
Particle Size 50 μm (nominal)
For Use With (Equipment) Robotic Fluid Handlers
For Use With (Application) Robotic Liquid Handling
Type RoboColumn
Diameter (Metric) Inner 0.5 cm
Content And Storage Store at 2°C to 8°C.
Product Line POROS CaptureSelect RoboColumn
Ligand Type CaptureSelect AAVX Affinity Ligand
Matrix Cross-linked poly(styrene-divinylbenzene) POROS beads
Target AAV
Column Format Pre-packed Robo Column
Bed Volume 600 μL
Show More Show Less