Learn More
Eisco™ Porcelain Spot Plate
Catalog No. S26588
Change view
Click to view available options
No. of Wells:
12
Catalog No. No. of Wells
S26588 12
1 options
Catalog No. S26588 Supplier Eisco™ Supplier No. CH0542B
Only null left
White porcelain spot plates are deal for observing reactions with color changes.
  • Spot plate made of porcelain
  • Available with six or twelve wells
  • White for good visibility of color change reactions
  • Perfect for any laboratory
Specifications
No. of Wells 12
Color White
Material Porcelain
Type Spot Plate