Learn More
Eisco™ Porcelain Desiccator Plates
Catalog No. S80820
Change view
Click to view available options
Diameter (Metric) Outer:
15 cm
20 cm
25 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Diameter (Metric) Outer
S80820 20 cm
S27043 15 cm
S27044 25 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S80820 Supplier Eisco™ Supplier No. CH0360B
Only null left
Plate has precisely cut out holes that are specifically designed for use in desiccator
  • Porcelain plate with precisely cut out holes
  • Designed for use in desiccator
  • An excellent choice for use in school laboratories
  • Autoclavable
Specifications
Diameter (Metric) Outer 20 cm
Product Type Desiccator Plate
Material Porcelain
Includes 7 Holes