missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Porcelain 25mL Crucibles

Catalog No. FB960S
Change view
Click to view available options
Product Type:
Fine
Medium
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
FB960S Medium
FB960R Fine
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. FB960S

For laboratory and industrial purpose

  • Material type: Porcelain
  • Color: White
  • Cubic feet/case: 0.237

Specifications

Color White
Description Pore Size: 15 microns
Material Porcelain
For Use With (Application) For laboratory and industrial purpose
Capacity (Metric) 25 mL
Product Type Medium