missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ Q, 100-120 mesh

Catalog No. 111004039
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004039 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004039 Supplier MilliporeSigma™ Supelco™ Supplier No. 20332

Porapak™™ Q packed in a stainless steel GC column maybe used for oxidation reduction experiments for designing of appropriate surface sites for ruthenium-ceria catalysts supported on graphite by controlled preparation method.

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. Porapak™™ Q is a porous polymer composed of ethylvynilbenzene and divynilbenzene mostly suitable for the separation of hydrocarbons, organic analytes in water, and oxides of nitrogen.

Specifications

Particle Size 100/120 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜550 m2/g
Product Line PoraPak
For Use With (Application) GC
Density ˜0.34 g/mL

PoraPak is a trademark of Waters