missing translation for 'onlineSavingsMsg'
Learn More

MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ P, 80-100 mesh

Catalog No. 111004037
Change view
Click to view available options
Quantity:
75 cc
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111004037 75 cc
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111004037 Supplier MilliporeSigma™ Supelco™ Supplier No. 20328

Stainless steel column packed with Porapak™™ P maybe used for the analysis of lipoxygenase reaction products by gas chromatography equipped with flame ionization detectors.

Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. Porapak™™ P is a porous polymer composed of styrene and divinylbenzene.

Specifications

Particle Size 80/100 Mesh
Packaging Bottle
Quantity 75 cc
Surface Area (Metric) ˜150 m2/g
Product Line PoraPak
For Use With (Application) GC
Density ˜0.26 g/mL (free fall density)

PoraPak is a trademark of Waters