missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MilliporeSigma™ Supelco™ Porapak™ Porous Polymer Adsorbent, Matrix Porapak™ P, 100-120 mesh
Description
Porous polymers are the most suitable adsorbent for applications based on the analysis of gases, acids, amines, organics of low carbon number and water. They are best suited for gas chromatography and are basically copolymers of polydivinylbenzene (DVB) which are very porous in nature ranging from mesoporous to microporous. Porapak™™ P is a porous polymer composed of styrene and divinylbenzene.
Specifications
Specifications
| Particle Size | 100/120 Mesh |
| Packaging | Bottle |
| Quantity | 75 cc |
| Surface Area (Metric) | ˜150 m2/g |
| Product Line | PoraPak |
| For Use With (Application) | GC |
| Density | ˜0.26 g/mL (free fall density) |
PoraPak is a trademark of Waters