missing translation for 'onlineSavingsMsg'
Learn More
Learn More
United Scientific™ Pop Beads
Description
- The beads connect easily and can be quickly disassembled to simulate different molecular activities.
- With these hands-on models, students can visually and tactually feel DNA models and simulate mitosis and meiosis.
- The specially designed white pop beads with four extra holes,equally spaced in a 90° plane from the axis of attachment, are typically used to form the nucleotide backbone. These are 10 mm diameter in size. The clear pop bead connectors are designed to simulate hydrogen bonds.
- Various colors to make a wide variety of models and kits
Caution: Choking Hazard: Children under 8 years can choke or suffocate. Adult supervision is required.
Specifications
Specifications
| Color | White |
| Height (Metric) | 3 mm |
| Material | Plastic |
| Quantity | 10 Pk. |
| Width (Metric) | 3 mm |
| Product Type | Pop Beads |